Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002965.3 | NP_002956.1 | 3 | 265 | ||
GenBank mRNA | AF086362.1 | - | 2 | 246 | ||
AK311882.1 | - | 3 | 266 | |||
BC047681.2 | - | 2 | 247 | 6060124 | ||
CR542207.1 | - | 2 | 222 | |||
CR542224.1 | - | 2 | 222 | |||
M26311.1 | - | 2 | 253 | |||
X06233.1 | - | 3 | 266 |