Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005622.3 | NP_005613.2 | 7 | 1186 | ||
NM_202000.2 | NP_973729.1 | 7 | 1186 | |||
XM_011545911.1 | XP_011544213.1 | 8 | 1291 | |||
XM_017023523.1 | XP_016879012.1 | 7 | 1180 | |||
XM_017023524.1 | XP_016879013.1 | 6 | 1050 | |||
GenBank mRNA | AK223139.1 | - | 7 | 1155 | ||
AK290322.1 | - | 7 | 1155 | |||
AK296122.1 | - | 7 | 985 | |||
BC002790.2 | - | 6 | 1045 | 3617826 | ||
BU170035.1 | - | 1 | 38 | |||
CA425752.1 | - | 3 | 399 | |||
D16350.1 | - | 6 | 1026 | |||
X80062.1 | - | 7 | 1135 |