Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291446.1 | NP_001278375.1 | 3 | 2365 | ||
NM_001291447.1 | NP_001278376.1 | 3 | 2345 | |||
NM_005407.2 | NP_005398.2 | 2 | 2770 | |||
XM_011537064.1 | XP_011535366.1 | 2 | 2696 | |||
XM_011537065.1 | XP_011535367.1 | 2 | 2842 | |||
GenBank mRNA | AB002358.2 | - | 2 | 2757 | ||
AF465630.1 | - | 1 | 855 | |||
AK293574.1 | - | 3 | 2051 | |||
AK294051.1 | - | 3 | 1840 | |||
AK296414.1 | - | 3 | 2164 | |||
AK299931.1 | - | 4 | 2049 | |||
BC136528.1 | - | 2 | 2505 | 9020518 | ||
BC136529.1 | - | 2 | 2505 | 9020519 | ||
X98834.1 | - | 2 | 2657 |