Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127892.1 | NP_001121364.1 | 2 | 1644 | ||
NM_002968.2 | NP_002959.2 | 2 | 1534 | |||
XM_006721241.3 | XP_006721304.1 | 3 | 1609 | |||
XM_011523254.2 | XP_011521556.1 | 3 | 1635 | |||
GenBank mRNA | AK295103.1 | - | 3 | 1424 | ||
AK307835.1 | - | 2 | 1644 | |||
EU668343.1 | - | 2 | 1503 | |||
Y18265.1 | - | 2 | 1503 |