Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303252.1 | NP_001290181.1 | 7 | 882 | ||
NM_002969.4 | NP_002960.2 | 8 | 912 | |||
GenBank mRNA | AK098058.1 | - | 2 | 870 | ||
AK123665.1 | - | 2 | 460 | |||
AK289770.1 | - | 4 | 616 | |||
AK294422.1 | - | 5 | 402 | |||
AK307421.1 | - | 6 | 703 | |||
AK307724.1 | - | 1 | 39 | |||
BC015741.1 | - | 8 | 813 | 4907996 | ||
BX640648.1 | - | 6 | 519 | |||
CB123561.1 | - | 7 | 485 | |||
CR456515.1 | - | 7 | 667 | |||
U66243.1 | - | 8 | 1064 | |||
X79483.1 | - | 8 | 695 | |||
Y10487.1 | - | 8 | 868 |