Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002970.3 | NP_002961.1 | 1 | 242 | ||
NR_027783.2 | 1 | 242 | ||||
GenBank mRNA | AF251292.1 | - | 1 | 253 | ||
AK310078.1 | - | 1 | 227 | |||
AK310094.1 | - | 1 | 223 | |||
AK312162.1 | - | 1 | 225 | |||
AK315888.1 | - | 1 | 165 | |||
AL050290.1 | - | 1 | 203 | |||
AY841998.1 | - | 1 | 49 | |||
BC002503.2 | - | 1 | 203 | 3051095 | ||
BC008424.1 | - | 1 | 227 | 4134501 | ||
BT006825.1 | - | 1 | 48 | |||
CD516700.1 | - | 1 | 248 | |||
CD638929.1 | - | 1 | 230 | |||
CN271682.1 | - | 1 | 242 | |||
CR450294.1 | - | 1 | 48 | |||
M55580.1 | - | 53 | ||||
M77693.1 | - | 1 | 213 |