Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002970.3 | NP_002961.1 | 1, 2 | 269 | ||
NR_027783.2 | 1, 2 | 269 | ||||
GenBank mRNA | AF251292.1 | - | 1, 2 | 280 | ||
AK310078.1 | - | 2 | 267 | |||
AK310094.1 | - | 2 | 461 | |||
AK312162.1 | - | 1, 2 | 252 | |||
AL050290.1 | - | 1, 2 | 230 | |||
AY841998.1 | - | 1, 2 | 76 | |||
BC002503.2 | - | 1, 2 | 230 | 3051095 | ||
BC008424.1 | - | 1, 2 | 254 | 4134501 | ||
BT006825.1 | - | 1, 2 | 75 | |||
CD516700.1 | - | 1, 2 | 275 | |||
CD638929.1 | - | 1, 2 | 257 | |||
CN271682.1 | - | 1, 2 | 269 | |||
CR450294.1 | - | 1, 2 | 75 | |||
M55580.1 | - | 80 | ||||
M77693.1 | - | 1, 2 | 240 |