Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005063.4 | NP_005054.3 | 5 | 1304 | ||
GenBank mRNA | AB032261.1 | - | 5 | 1194 | ||
AB208982.1 | - | 5 | 1061 | |||
AF097514.1 | - | 5 | 1049 | |||
AK312312.1 | - | 5 | 1086 | |||
BC005807.2 | - | 5 | 991 | |||
BC062303.1 | - | 5 | 1190 | 6586153 | ||
Y13647.1 | - | 5 | 1022 |