Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142349.1 | NP_001135821.1 | 2 | 488 | ||
NM_174934.3 | NP_777594.1 | 3 | 551 | |||
GenBank mRNA | AK314531.1 | - | 3 | 464 | ||
AY149967.1 | - | 3 | 462 | |||
BC035017.2 | - | 2 | 452 | 4817294 | ||
DA211092.1 | - | 2 | 452 |