Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002981.2 | NP_002972.1 | 1, 2 | 147 | ||
GenBank mRNA | AK312183.1 | - | 1, 2 | 145 | ||
BC105073.1 | - | 1, 2 | 112 | 8144076 | ||
BC105075.1 | - | 1, 2 | 112 | 8144078 | ||
DA620119.1 | - | 1, 2 | 145 | |||
M57502.1 | - | 1, 2 | 125 |