Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002981.2 | NP_002972.1 | 2 | 183 | ||
GenBank mRNA | AK312183.1 | - | 2 | 181 | ||
BC105073.1 | - | 2 | 148 | 8144076 | ||
BC105075.1 | - | 2 | 148 | 8144078 | ||
DA620119.1 | - | 2 | 181 | |||
M57502.1 | - | 2 | 161 |