Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002982.3 | NP_002973.1 | 2 | 174 | ||
GenBank mRNA | AV733621.2 | - | 2 | 174 | ||
BC009716.1 | - | 2 | 157 | 3901489 | ||
BT007329.1 | - | 2 | 101 | |||
M24545.1 | - | 2 | 170 | |||
M26683.1 | - | 1 | 39 | |||
S71513.1 | - | 2 | 154 | |||
X14768.1 | - | 2 | 154 |