Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004590.3 | NP_004581.1 | 3 | 328 | ||
XM_005258020.4 | XP_005258077.1 | 5 | 610 | |||
GenBank mRNA | AB007454.1 | - | 3 | 328 | ||
AF039955.1 | - | 3 | 319 | |||
AF055467.1 | - | 3 | 286 | |||
BC096352.4 | - | 4 | 524 | |||
BC099662.3 | - | 3 | 293 | 40009719 | ||
BC099663.1 | - | 4 | 524 | |||
BC099664.4 | - | 4 | 524 | |||
BQ722834.1 | - | 2 | 205 | |||
U91746.1 | - | 3 | 252 |