Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002998.3 | NP_002989.2 | 4 | 964 | ||
XM_011517212.2 | XP_011515514.1 | 5 | 496 | |||
GenBank mRNA | AK025488.1 | - | 4 | 805 | ||
AK074530.1 | - | 5 | 750 | |||
AK097839.1 | - | 5 | 765 | |||
AK130131.1 | - | 4 | 805 | |||
AK298643.1 | - | 4 | 664 | |||
AK298939.1 | - | 4 | 506 | |||
AK300067.1 | - | 4 | 702 | |||
BC030133.2 | - | 4 | 950 | 4138627 | ||
BC049836.1 | - | 4 | 682 | 4931549 | ||
J04621.1 | - | 4 | 934 |