Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000450.2 | NP_000441.2 | 6 | 1010 | ||
GenBank mRNA | AK314214.1 | - | 6 | 1010 | ||
BC131551.1 | - | 6 | 969 | 40085183 | ||
BC142677.1 | - | 6 | 997 | 40148214 | ||
BC142711.1 | - | 6 | 997 | 40148829 | ||
M24736.1 | - | 6 | 969 | |||
M30640.1 | - | 6 | 993 |