Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000655.4 | NP_000646.2 | 3 | 371 | ||
NR_029467.1 | 1 | 135 | ||||
GenBank mRNA | AB583235.1 | - | 2 | 167 | ||
AJ246000.1 | - | 3 | 242 | |||
AK225713.1 | - | 3 | 282 | |||
AK298705.1 | - | 1 | 130 | |||
AK312673.1 | - | 3 | 282 | |||
BC020758.1 | - | 3 | 268 | 4717456 | ||
DB271854.1 | - | 3 | 368 | |||
DB482055.1 | - | 3 | 365 | |||
DC378123.1 | - | 1 | 135 | |||
M25280.1 | - | 3 | 282 | |||
X16070.1 | - | 3 | 295 | |||
X16150.1 | - | 3 | 258 | |||
X17519.1 | - | 3 | 255 |