Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELPLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206609.1 | NP_001193538.1 | 2 | 362 | ||
NM_003006.4 | NP_002997.2 | 2 | 297 | |||
GenBank mRNA | AK127638.1 | - | 2 | 295 | ||
AK225644.1 | - | 2 | 193 | |||
AK290395.1 | - | 2 | 264 | |||
AK295238.1 | - | 2 | 199 | |||
AK298738.1 | - | 2 | 295 | |||
AK298742.1 | - | 2 | 334 | |||
AK300057.1 | - | 3 | 189 | |||
BC029782.1 | - | 2 | 276 | 5180824 | ||
DC339004.1 | - | 2 | 362 | |||
U02297.1 | - | 2 | 196 |