Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318798.1 | NP_001305727.1 | 18 | 1984 | ||
NM_001318800.1 | NP_001305729.1 | 18 | 2268 | |||
NM_004186.4 | NP_004177.3 | 19 | 2245 | |||
XM_005265381.4 | XP_005265438.1 | 19 | 2375 | |||
XM_005265382.4 | XP_005265439.1 | 18 | 2085 | |||
XM_006713289.3 | XP_006713352.1 | 13 | 1857 | |||
XM_011533998.2 | XP_011532300.1 | 19 | 2177 | |||
GenBank mRNA | AB209259.1 | - | 18 | 1938 | ||
BC042914.1 | - | 19 | 2241 | 5477898 | ||
U33920.1 | - | 18 | 2121 | |||
U38276.1 | - | 18 | 2262 |