Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005066.2 | NP_005057.1 | 4 | 1489 | ||
NR_136702.1 | 4 | 1492 | ||||
NR_136703.1 | 4 | 1492 | ||||
XM_005271112.4 | XP_005271169.1 | 4 | 1508 | |||
XM_005271113.4 | XP_005271170.1 | 4 | 1508 | |||
XM_005271115.4 | XP_005271172.1 | 4 | 1508 | |||
XM_011541950.2 | XP_011540252.1 | 4 | 1508 | |||
XM_017002053.1 | XP_016857542.1 | 4 | 1508 | |||
XM_017002054.1 | XP_016857543.1 | 4 | 1508 | |||
GenBank mRNA | BC004534.2 | - | 4 | 1177 | ||
BC027708.2 | - | 4 | 1480 | |||
BC051192.1 | - | 4 | 1491 | 5262885 | ||
X16850.1 | - | 3 | 463 | |||
X70944.1 | - | 4 | 1481 |