Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005066.2 | NP_005057.1 | 2 | 988 | ||
NR_136702.1 | 2 | 991 | ||||
NR_136703.1 | 2 | 991 | ||||
XM_005271112.4 | XP_005271169.1 | 2 | 1007 | |||
XM_005271113.4 | XP_005271170.1 | 2 | 1007 | |||
XM_005271115.4 | XP_005271172.1 | 2 | 1007 | |||
XM_011541950.2 | XP_011540252.1 | 2 | 1007 | |||
XM_017002053.1 | XP_016857542.1 | 2 | 1007 | |||
XM_017002054.1 | XP_016857543.1 | 2 | 1007 | |||
GenBank mRNA | AK307985.1 | - | 3 | 795 | ||
BC004534.2 | - | 2 | 676 | |||
BC027708.2 | - | 2 | 979 | |||
BC051192.1 | - | 2 | 990 | 5262885 | ||
X70944.1 | - | 2 | 980 |