Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003013.2 | NP_003004.1 | 3 | 890 | ||
GenBank mRNA | AF311912.1 | - | 3 | 886 | ||
AK075372.1 | - | 3 | 883 | |||
AK093922.1 | - | 4 | 858 | |||
AY359001.1 | - | 3 | 692 | |||
BC008666.1 | - | 3 | 870 | 3874262 |
Set Membership: |
G-protein coupled receptor |