Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000023.3 | NP_000014.1 | 6 | 748 | ||
NR_135553.1 | 6 | 748 | ||||
XM_011525120.2 | XP_011523422.2 | 6 | 1214 | |||
XM_011525122.2 | XP_011523424.2 | 6 | 1214 | |||
XM_011525124.2 | XP_011523426.1 | 5 | 569 | |||
XM_017024950.1 | XP_016880439.1 | 6 | 1210 | |||
GenBank mRNA | AK290622.1 | - | 6 | 721 | ||
AK293614.1 | - | 5 | 569 | |||
AK300710.1 | - | 6 | 730 | |||
BC025702.1 | - | 6 | 720 | 5222483 | ||
KU178331.1 | - | 6 | 692 | |||
L34355.1 | - | 6 | 729 | |||
U08895.1 | - | 6 | 703 |