Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 12, 13 | 1930 | ||
NM_001143677.1 | NP_001137149.1 | 10, 11 | 1181 | |||
NM_001143678.1 | NP_001137150.1 | 10, 11 | 1360 | |||
NM_001291995.1 | NP_001278924.1 | 9, 10 | 1004 | |||
NM_005627.3 | NP_005618.2 | 10, 11 | 1136 | |||
XM_011536071.1 | XP_011534373.1 | 13, 14 | 1712 | |||
GenBank mRNA | AF153609.1 | - | 10, 11 | 1089 | ||
AK055077.1 | - | 12, 13 | 1930 | |||
AK098509.1 | - | 10, 11 | 1104 | |||
AK293383.1 | - | 10, 11 | 1170 | |||
AK295422.1 | - | 9, 10 | 972 | |||
AK298688.1 | - | 10, 11 | 1419 | |||
BC001263.1 | - | 10, 11 | 1089 | 3459056 | ||
EU518415.1 | - | 12, 13 | 1332 | |||
FM205707.1 | - | 10, 11 | 1124 | |||
FM205708.1 | - | 10, 11 | 1159 | |||
FM205709.1 | - | 12, 13 | 1779 | |||
FM205710.1 | - | 9, 10 | 1248 | |||
Y10032.1 | - | 10, 11 | 1089 |