Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291995.1 | NP_001278924.1 | 1, 2 | 155 | ||
NM_005627.3 | NP_005618.2 | 1, 2 | 155 | |||
GenBank mRNA | AF153609.1 | - | 1, 2 | 108 | ||
AK098509.1 | - | 1, 2 | 123 | |||
AK295422.1 | - | 1, 2 | 123 | |||
BC001263.1 | - | 1, 2 | 108 | 3459056 | ||
BX649005.1 | - | 1, 2 | 126 | |||
DB450096.1 | - | 1, 2 | 155 | |||
Y10032.1 | - | 1, 2 | 108 |