Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144757.1 | NP_001138229.1 | 2 | 142 | ||
NM_003020.3 | NP_003011.1 | 2 | 142 | |||
GenBank mRNA | AK314965.1 | - | 2 | 135 | ||
BC005349.1 | - | 2 | 81 | 3949961 | ||
BC093053.1 | - | 2 | 101 | 30720075 | ||
BP377248.1 | - | 2 | 142 | |||
CR541824.1 | - | 1 | 25 | |||
Y00757.1 | - | 2 | 53 |