Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003021.3 | NP_003012.1 | 3 | 319 | ||
XM_011528178.2 | XP_011526480.1 | 3 | 373 | |||
GenBank mRNA | AF368279.1 | - | 2 | 193 | ||
AF408399.1 | - | 2 | 172 | |||
AJ133129.1 | - | 3 | 214 | |||
AJ223828.1 | - | 2 | 213 | |||
AK293535.1 | - | 3 | 290 | |||
AL050156.1 | - | 3 | 281 | |||
BC000390.2 | - | 3 | 222 | 2821268 | ||
BC002989.1 | - | 3 | 222 | 2821268 | ||
BC005165.2 | - | 3 | 216 | 3532317 | ||
BC008885.2 | - | 3 | 216 | 4302652 | ||
BG423904.1 | - | 320 | ||||
CR542282.1 | - | 2 | 172 |