Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122681.1 | NP_001116153.1 | 5 | 509 | ||
NM_001145855.1 | NP_001139327.1 | 5 | 652 | |||
NM_001145856.1 | NP_001139328.1 | 5 | 599 | |||
NM_003023.4 | NP_003014.3 | 5 | 650 | |||
GenBank mRNA | AB000462.1 | - | 5 | 650 | ||
AB000463.1 | - | 6 | 829 | |||
AF000936.1 | - | 4 | 389 | |||
AK225497.1 | - | 5 | 511 | |||
AK299996.1 | - | 5 | 568 | |||
AK304752.1 | - | 4 | 949 | |||
AK312286.1 | - | 5 | 447 | |||
BC022996.1 | - | 5 | 505 | 5103986 | ||
U56386.1 | - | 5 | 597 |