Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003028.2 | NP_003019.2 | 2, 3 | 1397 | ||
GenBank mRNA | BC136581.1 | - | 2, 3 | 1270 | 9020571 | |
BC136582.1 | - | 2, 3 | 1270 | 9020572 | ||
X75342.1 | - | 2, 3 | 1400 |