Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254757.1 | NP_001241686.1 | 5 | 480 | ||
NM_001254758.1 | NP_001241687.1 | 5 | 353 | |||
NM_001254759.1 | NP_001241688.1 | 3 | 497 | |||
NM_006278.2 | NP_006269.1 | 5 | 679 | |||
XM_011542959.1 | XP_011541261.1 | 4 | 282 | |||
XM_011542960.2 | XP_011541262.1 | 4 | 282 | |||
XM_017018134.1 | XP_016873623.1 | 5 | 365 | |||
GenBank mRNA | AF035249.1 | - | 1 | 49 | ||
AF516602.1 | - | 3 | 207 | |||
AF516603.1 | - | 3 | 177 | |||
AF516604.1 | - | 3 | 207 | |||
AF525084.1 | - | 3 | 237 | |||
AK021929.1 | - | 3 | 181 | |||
AK291577.1 | - | 4 | 309 | |||
AK314738.1 | - | 4 | 279 | |||
AL520639.3 | - | 5 | 438 | |||
AL529887.3 | - | 5 | 354 | |||
AY040826.1 | - | 4 | 240 | |||
BC010645.1 | - | 5 | 353 | 3449625 | ||
BI259855.1 | - | 5 | 448 | |||
CR456858.1 | - | 4 | 210 | |||
DA210431.1 | - | 5 | 480 | |||
DA841563.1 | - | 3 | 498 | |||
L23767.1 | - | 3 | 219 | |||
X74570.1 | - | 5 | 372 |