Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304450.1 | NP_001291379.1 | 1, 2 | 713 | ||
NM_003034.3 | NP_003025.1 | 1, 2 | 713 | |||
GenBank mRNA | AK290941.1 | - | 1, 2 | 354 | ||
AK297310.1 | - | 1, 2 | 196 | |||
AK315340.1 | - | 1, 2 | 688 | |||
AY569975.1 | - | 1, 2 | 237 | |||
BC117285.1 | - | 1, 2 | 632 | 40125836 | ||
BC126162.1 | - | 1, 2 | 632 | 8991878 | ||
D26360.1 | - | 1, 2 | 334 | |||
EU041716.1 | - | 1, 2 | 713 | |||
L32867.1 | - | 1, 2 | 298 | |||
L43494.1 | - | 1, 2 | 242 | |||
X77922.1 | - | 1, 2 | 713 |