Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200053.1 | NP_001186982.1 | 4 | 1010 | ||
NM_001200054.1 | NP_001186983.1 | 6 | 1268 | |||
NM_006928.4 | NP_008859.1 | 7 | 1303 | |||
XM_006719569.1 | XP_006719632.1 | 6 | 1880 | |||
XM_011538685.1 | XP_011536987.1 | 7 | 1332 | |||
GenBank mRNA | AK092881.1 | - | 6 | 1881 | ||
AK130180.1 | - | 7 | 1327 | |||
AK300150.1 | - | 4 | 990 | |||
BC001414.2 | - | 6 | 1227 | 3139788 | ||
BT007202.1 | - | 6 | 1217 | |||
M32295.1 | - | 6 | 1238 | |||
M77348.1 | - | 6 | 1228 | |||
S73003.1 | - | 6 | 1238 | |||
U01874.1 | - | 6 | 1223 |