Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004170.5 | NP_004161.4 | 9 | 1230 | ||
XM_011518007.1 | XP_011516309.1 | 10 | 1250 | |||
XM_011518008.2 | XP_011516310.1 | 10 | 8495 | |||
XM_011518009.2 | XP_011516311.1 | 9 | 8425 | |||
XM_011518010.1 | XP_011516312.1 | 8 | 1040 | |||
XM_017015042.1 | XP_016870531.1 | 9 | 1142 | |||
XM_017015043.1 | XP_016870532.1 | 8 | 1083 | |||
GenBank mRNA | AB008536.1 | - | 9 | 1168 | ||
AF037982.1 | - | 2 | 176 | |||
AK313013.1 | - | 9 | 1131 | |||
BC033040.1 | - | 9 | 1216 | 5261168 | ||
U03506.1 | - | 9 | 995 | |||
U06469.1 | - | 9 | 1016 | |||
U08989.1 | - | 9 | 1142 |