Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272087.1 | NP_001259016.1 | 4 | 1576 | ||
NM_001272088.1 | NP_001259017.1 | 2 | 404 | |||
NM_005071.2 | NP_005062.1 | 1 | 182 | |||
NR_073589.1 | 2 | 340 | ||||
XM_006722842.1 | XP_006722905.1 | 2 | 365 | |||
XM_006722844.1 | XP_006722907.1 | 2 | 258 | |||
XM_017027152.1 | XP_016882641.1 | 2 | 453 | |||
GenBank mRNA | AB209199.1 | - | 1 | 3343 | ||
AK302380.1 | - | 2 | 317 | |||
AK312996.1 | - | 2 | 448 | |||
BC028721.2 | - | 2 | 326 | 5269300 | ||
BC040604.1 | - | 4 | 1531 | 5270965 | ||
BI829415.1 | - | 2 | 292 | |||
DA200776.1 | - | 2 | 493 | |||
DB027852.1 | - | 2 | 340 | |||
DR004792.1 | - | 2 | 390 | |||
U18244.1 | - | 1 | 182 |