Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000341.3 | NP_000332.2 | 7 | 1337 | ||
XM_011533047.2 | XP_011531349.1 | 7 | 1337 | |||
GenBank mRNA | AB033549.1 | - | 7 | 1318 | ||
AK091932.1 | - | 8 | 1203 | |||
AK092000.1 | - | 4 | 1443 | |||
AK223146.1 | - | 7 | 1299 | |||
AK289636.1 | - | 7 | 1315 | |||
AK309145.1 | - | 7 | 1331 | |||
AK313157.1 | - | 7 | 1331 | |||
BC022386.1 | - | 7 | 1335 | 4556946 | ||
BC093624.1 | - | 7 | 1296 | 7939469 | ||
BC093626.1 | - | 7 | 1291 | 7939471 | ||
DQ023512.1 | - | 4 | 1443 | |||
DQ023514.1 | - | 1 | 460 | |||
DQ023515.1 | - | 7 | 1259 | |||
DQ023516.1 | - | 7 | 1259 | |||
DQ023517.1 | - | 7 | 1259 | |||
KU178335.1 | - | 7 | 1259 | |||
L11696.1 | - | 7 | 1302 | |||
M95548.1 | - | 7 | 1303 |