Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000343.3 | NP_000334.1 | 15 | 2185 | ||
NM_001256314.1 | NP_001243243.1 | 14 | 1903 | |||
GenBank mRNA | AK297665.1 | - | 14 | 1877 | ||
AK312948.1 | - | 15 | 2089 | |||
AK316184.1 | - | 14 | 1882 | |||
BC139839.1 | - | 15 | 1954 | |||
CR456579.1 | - | 15 | 1985 | |||
M24847.1 | - | 15 | 1945 |