Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045.5 | NP_001036.1 | 1856 | |||
GenBank mRNA | AK308014.1 | - | 10 | 1587 | ||
AK309538.1 | - | 1 | 24 | |||
AK313166.1 | - | 10 | 1585 | |||
AY902473.1 | - | 8 | 1351 | |||
BC069484.1 | - | 8 | 1328 | 7262380 | ||
L05568.1 | - | 8 | 1352 | |||
X70697.1 | - | 8 | 1390 |