Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC6A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045.5 | NP_001036.1 | 1064 | |||
GenBank mRNA | AK308014.1 | - | 4, 5 | 795 | ||
AK313166.1 | - | 4, 5 | 793 | |||
AY902473.1 | - | 2, 3 | 559 | |||
BC069484.1 | - | 2, 3 | 536 | 7262380 | ||
BX399758.2 | - | 4, 5 | 823 | |||
L05568.1 | - | 2, 3 | 560 | |||
X70697.1 | - | 2, 3 | 598 |