Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323971.1 | NP_001310900.1 | 7 | 1434 | ||
NM_001323972.1 | NP_001310901.1 | 6 | 1131 | |||
NM_001323973.1 | NP_001310902.1 | 7 | 1434 | |||
NM_001323974.1 | NP_001310903.1 | 7 | 1348 | |||
NM_001323975.1 | NP_001310904.1 | 7 | 1348 | |||
NM_004594.2 | NP_004585.1 | 7 | 1354 | |||
NR_136664.1 | 7 | 1348 | ||||
NR_136665.1 | 7 | 1295 | ||||
NR_136666.1 | 6 | 1256 | ||||
NR_136667.1 | 7 | 1348 | ||||
XM_017023594.1 | XP_016879083.1 | 7 | 2757 | |||
GenBank mRNA | AB209203.1 | - | 6 | 2159 | ||
AF111173.1 | - | 7 | 1360 | |||
BC142671.1 | - | 7 | 1317 | 40148180 | ||
BC144131.1 | - | 7 | 1295 | |||
BC150207.1 | - | 6 | 1610 |