Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166496.1 | NP_001159968.1 | 5 | 2115 | ||
NM_003051.3 | NP_003042.3 | 5 | 1646 | |||
XM_011542026.2 | XP_011540328.1 | 5 | 1379 | |||
XM_011542027.2 | XP_011540329.1 | 5 | 1389 | |||
GenBank mRNA | AK000641.1 | - | 2 | 531 | ||
AK094064.1 | - | 5 | 2115 | |||
AK296688.1 | - | 5 | 1429 | |||
AK303603.1 | - | 5 | 1488 | |||
AK312500.1 | - | 5 | 1489 | |||
AL162079.1 | - | 5 | 1476 | |||
BC026317.1 | - | 5 | 1489 | 4811581 | ||
L31801.1 | - | 4 | 1295 |