Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135691.2 | NP_001129163.1 | 16, 17 | 1927 | ||
NM_001142324.1 | NP_001135796.1 | 15, 16 | 1831 | |||
NM_003053.3 | NP_003044.1 | 15, 16 | 1723 | |||
XM_011544625.1 | XP_011542927.1 | 14, 15 | 1572 | |||
XM_011544626.1 | XP_011542928.1 | 14, 15 | 1560 | |||
GenBank mRNA | AK313720.1 | - | 15, 16 | 1619 | ||
BC006317.2 | - | 15, 16 | 1740 | 4125784 | ||
BC009387.2 | - | 16, 17 | 1744 | 4127678 | ||
U39905.1 | - | 15, 16 | 1723 |