Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC20A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257180.1 | NP_001244109.1 | 2 | 780 | ||
NM_001257181.1 | NP_001244110.1 | 2 | 411 | |||
NM_006749.4 | NP_006740.1 | 2 | 743 | |||
XM_005273613.3 | XP_005273670.1 | 2 | 518 | |||
XM_005273615.3 | XP_005273672.1 | 2 | 518 | |||
XM_006716390.3 | XP_006716453.1 | 2 | 518 | |||
XM_006716391.3 | XP_006716454.1 | 2 | 3863 | |||
XM_017013748.1 | XP_016869237.1 | 2 | 634 | |||
XM_017013749.1 | XP_016869238.1 | 2 | 3863 | |||
XM_017013750.1 | XP_016869239.1 | 2 | 518 | |||
XM_017013751.1 | XP_016869240.1 | 2 | 392 | |||
XM_017013752.1 | XP_016869241.1 | 2 | 634 | |||
GenBank mRNA | AK291202.1 | - | 2 | 780 | ||
AK314774.1 | - | 2 | 412 | |||
BC028600.2 | - | 2 | 455 | 4820601 | ||
L20852.1 | - | 1 | 329 |