Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021094.3 | NP_066580.1 | 2 | 420 | ||
NM_134431.3 | NP_602307.1 | 4 | 890 | |||
XM_005253474.3 | XP_005253531.1 | 3 | 311 | |||
XM_005253477.3 | XP_005253534.1 | 2 | 198 | |||
XM_011520818.1 | XP_011519120.1 | 3 | 318 | |||
XM_011520819.1 | XP_011519121.1 | 3 | 465 | |||
XM_011520820.2 | XP_011519122.1 | 3 | 1787 | |||
XM_017019849.1 | XP_016875338.1 | 3 | 318 | |||
XM_017019850.1 | XP_016875339.1 | 2 | 198 | |||
GenBank mRNA | AF085224.1 | - | 3 | 379 | ||
AF279784.1 | - | 4 | 890 | |||
BC042452.1 | - | 2 | 200 | 4820050 | ||
DA216595.1 | - | 3 | 322 | |||
U21943.1 | - | 2 | 222 |