Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 28 | 4205 | ||
NM_001289397.1 | NP_001276326.1 | 28 | 4044 | |||
NM_001289398.1 | NP_001276327.1 | 2 | 226 | |||
NM_001289399.1 | NP_001276328.1 | 3 | 213 | |||
NM_001289400.1 | NP_001276329.1 | 3 | 634 | |||
NM_003070.4 | NP_003061.3 | 28 | 4218 | |||
NM_139045.3 | NP_620614.2 | 28 | 4218 | |||
GenBank mRNA | AK296373.1 | - | 28 | 3606 | ||
AK298045.1 | - | 3 | 213 | |||
AK299683.1 | - | 19 | 2411 | |||
AK300093.1 | - | 4 | 278 | |||
AK308941.1 | - | 2 | 371 | |||
AL558829.3 | - | 3 | 165 | |||
BI758293.1 | - | 2 | 227 | |||
CD513892.1 | - | 1 | 22 | |||
CD702931.1 | - | 2 | 465 | |||
DA084762.1 | - | 2 | 223 | |||
DA823500.1 | - | 2 | 148 | |||
X72889.1 | - | 28 | 4206 |