Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_023068.3 | NP_075556.1 | 5 | 1167 | ||
XM_006723610.3 | XP_006723673.1 | 6 | 1362 | |||
XM_011529324.2 | XP_011527626.1 | 6 | 1359 | |||
XM_011529325.2 | XP_011527627.1 | 6 | 1365 | |||
XM_011529326.2 | XP_011527628.1 | 6 | 1361 | |||
XM_011529327.2 | XP_011527629.1 | 5 | 1295 | |||
XM_011529328.2 | XP_011527630.1 | 6 | 1307 | |||
XM_011529329.1 | XP_011527631.1 | 5 | 2548 | |||
XM_011529330.1 | XP_011527632.1 | 5 | 2548 | |||
XM_011529331.1 | XP_011527633.1 | 5 | 2466 | |||
XM_011529332.1 | XP_011527634.1 | 4 | 752 | |||
XM_011529333.2 | XP_011527635.1 | 5 | 2548 | |||
GenBank mRNA | AF230073.1 | - | 5 | 1167 | ||
AY927519.1 | - | LIT3529 AY927519 |
4 | 1075 | ||
BC141884.1 | - | 5 | 1167 |