Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278656.1 | NP_001265585.1 | 3 | 613 | ||
NM_004175.4 | NP_004166.1 | 3 | 627 | |||
GenBank mRNA | AB451249.1 | - | 2 | 187 | ||
AB451373.1 | - | 2 | 187 | |||
AK296173.1 | - | 3 | 270 | |||
BC000457.2 | - | 3 | 283 | 2822042 | ||
BC003150.1 | - | 3 | 283 | 2822042 | ||
BC034447.1 | - | 3 | 310 | |||
BF344644.1 | - | 3 | 621 | |||
BM928709.1 | - | 3 | 575 | |||
BY993733.1 | - | 3 | 299 | |||
CN286602.1 | - | 3 | 267 | |||
CR456583.1 | - | 3 | 303 | |||
U15009.1 | - | 3 | 274 |