Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242933.1 | NP_001229862.1 | 6 | 728 | ||
NM_003099.4 | NP_003090.2 | 6 | 728 | |||
NM_148955.3 | NP_683758.1 | 4 | 533 | |||
XM_005254611.2 | XP_005254668.1 | 6 | 704 | |||
GenBank mRNA | AB209013.1 | - | 6 | 535 | ||
AF065483.1 | - | 6 | 598 | |||
AF065484.1 | - | 4 | 403 | |||
AK128179.1 | - | 6 | 620 | |||
AK222793.1 | - | 6 | 601 | |||
AK291752.1 | - | 6 | 612 | |||
AK310618.1 | - | 3 | 369 | |||
BC000357.2 | - | 6 | 608 | 2964409 | ||
BQ935515.1 | - | 6 | 602 | |||
BT006983.1 | - | 6 | 598 | |||
U53225.1 | - | 6 | 643 |