Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242933.1 | NP_001229862.1 | 10 | 1103 | ||
NM_003099.4 | NP_003090.2 | 10 | 1103 | |||
NM_148955.3 | NP_683758.1 | 8 | 908 | |||
XM_005254611.2 | XP_005254668.1 | 10 | 1079 | |||
GenBank mRNA | AB209013.1 | - | 10 | 910 | ||
AF065483.1 | - | 10 | 973 | |||
AF065484.1 | - | 8 | 778 | |||
AK128179.1 | - | 10 | 995 | |||
AK222561.1 | - | 10 | 987 | |||
AK222793.1 | - | 10 | 976 | |||
AK291752.1 | - | 10 | 987 | |||
AK310618.1 | - | 7 | 787 | |||
BC000357.2 | - | 10 | 983 | 2964409 | ||
BT006983.1 | - | 10 | 973 | |||
U53225.1 | - | 10 | 1018 |