Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005633.3 | NP_005624.2 | 10 | 1752 | ||
XM_005264515.4 | XP_005264572.1 | 10 | 1981 | |||
XM_011533062.2 | XP_011531364.1 | 10 | 1920 | |||
XM_011533064.2 | XP_011531366.1 | 10 | 1660 | |||
XM_011533066.2 | XP_011531368.1 | 3 | 661 | |||
GenBank mRNA | AB209140.1 | - | 3 | 723 | ||
AK290228.1 | - | 11 | 1798 | |||
L13857.1 | - | 10 | 1711 |