Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324113.1 | NP_001311042.1 | 5 | 826 | ||
NM_001324114.1 | NP_001311043.1 | 4 | 714 | |||
NM_001324115.1 | NP_001311044.1 | 5 | 1053 | |||
NM_001324116.1 | NP_001311045.1 | 4 | 941 | |||
NM_001324117.1 | NP_001311046.1 | 4 | 941 | |||
NM_003115.4 | NP_003106.3 | 4 | 875 | |||
XM_011509913.2 | XP_011508215.1 | 5 | 774 | |||
XM_011509914.2 | XP_011508216.1 | 5 | 826 | |||
GenBank mRNA | AB011004.1 | - | 3 | 546 | ||
AK312685.1 | - | 4 | 801 | |||
BC009377.2 | - | 4 | 848 | 4121439 | ||
S73498.1 | - | 4 | 778 |