Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130438.2 | NP_001123910.1 | 55 | 7180 | ||
NM_001195532.1 | NP_001182461.1 | 53 | 7105 | |||
NM_003127.3 | NP_003118.2 | 54 | 7165 | |||
XM_006717245.1 | XP_006717308.1 | 56 | 7150 | |||
XM_006717246.1 | XP_006717309.1 | 55 | 7135 | |||
XM_006717247.2 | XP_006717310.1 | 55 | 7090 | |||
XM_006717248.2 | XP_006717311.1 | 55 | 7086 | |||
XM_006717249.1 | XP_006717312.1 | 54 | 7072 | |||
XM_006717250.1 | XP_006717313.1 | 54 | 7069 | |||
XM_006717251.2 | XP_006717314.1 | 55 | 7177 | |||
XM_006717252.2 | XP_006717315.1 | 54 | 7027 | |||
XM_006717253.1 | XP_006717316.1 | 53 | 7012 | |||
XM_006717254.2 | XP_006717317.1 | 56 | 7237 | |||
XM_017015059.1 | XP_016870548.1 | 54 | 7147 | |||
XM_017015060.1 | XP_016870549.1 | 53 | 7009 | |||
XM_017015061.1 | XP_016870550.1 | 54 | 7085 | |||
GenBank mRNA | AB191262.1 | - | 53 | 7070 | ||
AB209860.1 | - | 54 | 7127 | |||
AK126576.1 | - | 4 | 992 | |||
AL110273.1 | - | 33 | 3992 | |||
BC053521.1 | - | 53 | 7054 | 5529391 | ||
J05243.1 | - | 54 | 7125 | |||
U83867.1 | - | 55 | 7107 | |||
X86901.1 | - | 8 | 849 |